accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1555-94-8

IUPAC Name: 8-ethoxyquinoline
Synonyms: 8-ethoxyquinoline
Molecular Weight (g/mol): 173.22
Molecular Formula: C11H11NO
InChi Key: UGRGZOJMYMJWLZ-UHFFFAOYSA-N
SMILES: CCOC1=C2N=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results