Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Ethoxyquinoline, 99%, Thermo Scientific™

Catalog No. AAA1699714
Change view
Click to view available options
Quantity:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1699714 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1699714 Supplier Thermo Scientific Chemicals Supplier No. A1699714

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1555-94-8
Molecular Formula C11H11NO
Quantity 25g
MDL Number MFCD00039728
Synonym quinoline, 8-ethoxy, ccris 7822, acmc-1byfx, 8-ethoxy quinoline
InChI Key UGRGZOJMYMJWLZ-UHFFFAOYSA-N
SMILES CCOC1=CC=CC2=C1N=CC=C2
IUPAC Name 8-ethoxyquinoline
Molecular Weight (g/mol) 173.215
PubChem CID 73781
Formula Weight 173.21
Percent Purity 99%
Chemical Name or Material 8-Ethoxyquinoline
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.