accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 155684-30-3

IUPAC Name: trimethoxy(2-{7-oxabicyclo[4.1.0]heptan-3-yl}ethyl)silane
Synonyms: trimethoxy(2-{7-oxabicyclo[4.1.0]heptan-3-yl}ethyl)silane
Molecular Weight (g/mol): 246.38
Molecular Formula: C11H22O4Si
InChi Key: DQZNLOXENNXVAD-UHFFFAOYNA-N
SMILES: CO[Si](CCC1CCC2OC2C1)(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results