Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β(3,4-Epoxycyclohexyl)-ethyltrimethoxysilane, MP Biomedicals

Catalog No. ICN224487 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN224487 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN224487 Supplier MP Biomedicals, Inc Supplier No. 0522448725

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 155684-30-3
Molecular Formula C11H22O4Si
Molecular Weight (g/mol) 246.378
InChI Key DQZNLOXENNXVAD-UHFFFAOYSA-N
Synonym 2-3,4-epoxycyclohexyl ethyltrimethoxysilane, silicone a-186, 2-7-oxabicyclo 4.1.0 heptan-3-yl ethyl trimethoxysilane, silane a 186, union carbide a-186, silane y-4086, a 186 heterocycle, silquest a 186, nuca 186, sila-ace s 530
PubChem CID 18819
IUPAC Name trimethoxy-[2-(7-oxabicyclo[4.1.0]heptan-4-yl)ethyl]silane
SMILES CO[Si](CCC1CCC2C(C1)O2)(OC)OC

Specifications

Quantity 100 g
Formula Weight 246.3
Chemical Name or Material Beta-(3, 4-Epoxycyclohexyl)- Ethyltrimethoxysilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.