accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFBrF

CAS RN 1559-88-2

IUPAC Name: 3-bromo-1,2,4,5-tetrafluorobenzene
Synonyms: 3-bromo-1,2,4,5-tetrafluorobenzene
Molecular Weight (g/mol): 228.97
Molecular Formula: C6HBrF4
InChi Key: YHAFCGSUIAFUCX-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(F)C(Br)=C1F
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)