Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2,3,5,6-tetrafluorobenzene, 98%

Catalog No. AAB2426514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2426514 25 g
AAB2426506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2426514 Supplier Thermo Scientific Chemicals Supplier No. B2426514
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1559-88-2 | C6HBrF4 | 228.97 g/mol

1-Bromo-2,3,5,6-tetrafluorobenzene is used to produce 1,2,4,5-tetrafluoro-3-hept-1-ynyl-benzene. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2,3,5,6-tetrafluorobenzene is used to produce 1,2,4,5-tetrafluoro-3-hept-1-ynyl-benzene. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1559-88-2
Molecular Formula C6HBrF4
Molecular Weight (g/mol) 228.97
MDL Number MFCD00000308
InChI Key YHAFCGSUIAFUCX-UHFFFAOYSA-N
Synonym 1-bromo-2,3,5,6-tetrafluorobenzene, 2,3,5,6-tetrafluorobromobenzene, bromo-2,3,5,6-tetrafluorobenzene, 1-bromo-2,3,5,6-tetrafluorbenzene, benzene, 3-bromo-1,2,4,5-tetrafluoro, 2,3,5,6-tetrafluoro bromobenzene, 2,3,5,6-tetrafluorobromo benzene, 1,2,4,5-tetrafluoro-3-bromobenzene, pubchem2196, acmc-1bwvg
PubChem CID 73794
IUPAC Name 3-bromo-1,2,4,5-tetrafluorobenzene
SMILES FC1=CC(F)=C(F)C(Br)=C1F

Specifications

Density 1.883
Boiling Point 142°C to 144°C
Flash Point 53°C (129°F)
Odor Aromatic
Refractive Index 1.469
Quantity 25 g
UN Number UN1993
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 228.98
Percent Purity 99%
Chemical Name or Material 1-Bromo-2,3,5,6-tetrafluorobenzene
Show More Show Less
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 216-327-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.