Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2,3,5,6-tetrafluorobenzene 97.0+%, TCI America™
SDP

Catalog No. B17065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17065G 5 g
B170625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17065G Supplier TCI America Supplier No. B17065G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1559-88-2
Molecular Formula C6HBrF4
Molecular Weight (g/mol) 228.97
MDL Number MFCD00000308
InChI Key YHAFCGSUIAFUCX-UHFFFAOYSA-N
Synonym 1-bromo-2,3,5,6-tetrafluorobenzene, 2,3,5,6-tetrafluorobromobenzene, bromo-2,3,5,6-tetrafluorobenzene, 1-bromo-2,3,5,6-tetrafluorbenzene, benzene, 3-bromo-1,2,4,5-tetrafluoro, 2,3,5,6-tetrafluoro bromobenzene, 2,3,5,6-tetrafluorobromo benzene, 1,2,4,5-tetrafluoro-3-bromobenzene, pubchem2196, acmc-1bwvg
PubChem CID 73794
IUPAC Name 3-bromo-1,2,4,5-tetrafluorobenzene
SMILES FC1=CC(F)=C(F)C(Br)=C1F

Specifications

Color Colorless
Boiling Point 143°C
Quantity 5 g
UN Number 1993
Formula Weight 228.97
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2,3,5,6-tetrafluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.