CAS RN 155906-10-8
IUPAC Name:
2-bromo-1,3-difluoro-5-iodobenzene
Synonyms:
2-bromo-1,3-difluoro-5-iodobenzene
Molecular Weight (g/mol):
318.89
Molecular Formula:
C6H2BrF2I
InChi Key:
WOLGNRYEQPISPB-UHFFFAOYSA-N
SMILES:
FC1=CC(I)=CC(F)=C1Br
1
–
1
of
1
results
1
2-Bromo-1,3-difluoro-5-iodobenzene 98.0+%, TCI America™
CAS: 155906-10-8 Molecular Formula: C6H2BrF2I Molecular Weight (g/mol): 318.89 MDL Number: MFCD06797752 InChI Key: WOLGNRYEQPISPB-UHFFFAOYSA-N PubChem CID: 2761406 IUPAC Name: 2-bromo-1,3-difluoro-5-iodobenzene SMILES: FC1=CC(I)=CC(F)=C1Br