accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15596-07-3

PCCO

CAS RN 15596-07-3

IUPAC Name: (triphenyl-λ⁵-phosphanylidene)ethenone
Synonyms: (triphenyl-λ⁵-phosphanylidene)ethenone
Molecular Weight (g/mol): 302.31
Molecular Formula: C20H15OP
InChi Key: MNASRBWCHRURHY-UHFFFAOYSA-N
SMILES: O=C=C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results