CAS RN 156006-28-9
IUPAC Name:
magnesium(2+) 3,4,5-trifluorobenzen-1-ide bromide
Synonyms:
magnesium(2+) 3,4,5-trifluorobenzen-1-ide bromide
Molecular Weight (g/mol):
235.29
Molecular Formula:
C6H2BrF3Mg
InChi Key:
FAANOBAJWDSCGM-UHFFFAOYSA-M
SMILES:
[Mg++].[Br-].FC1=C[C-]=CC(F)=C1F
1
–
1
of
1
results
1
Sigma Aldrich 3,5-Difluorophenylmagnesium Bromide Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.