accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15646-46-5

IUPAC Name: (4Z)-4-(ethoxymethylidene)-2-phenyl-4,5-dihydro-1,3-oxazol-5-one
Synonyms: (4Z)-4-(ethoxymethylidene)-2-phenyl-1,3-oxazol-5-one
Molecular Weight (g/mol): 217.22
Molecular Formula: C12H11NO3
InChi Key: SJHPCNCNNSSLPL-NTMALXAHSA-N
SMILES: CCO\C=C1/N=C(OC1=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)
Industry Type 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)