CAS RN 156840-13-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ HBC 530
GFP fluorophore mimic for imaging RNA in live cells; activated by binding to Pepper aptamers; suitable for confocal and two-photon microscopy