CAS RN 15687-27-1
Ibuprofen, USP grade, MP Biomedicals™
CAS: 15687-27-1 Molecular Formula: C13H18O2 Molecular Weight (g/mol): 206.29 MDL Number: MFCD00010393 InChI Key: HEFNNWSXXWATRW-UHFFFAOYNA-N Synonym: ibuprofen,2-4-isobutylphenyl propanoic acid,motrin,brufen,nurofen,advil,dolgit,liptan,nuprin,anflagen PubChem CID: 3672 ChEBI: CHEBI:5855 IUPAC Name: 2-[4-(2-methylpropyl)phenyl]propanoic acid SMILES: CC(C)CC1=CC=C(C=C1)C(C)C(O)=O
2-(4-Isobutylphenyl)propionic Acid 98.0+%, TCI America™
CAS: 15687-27-1 Molecular Formula: C13H18O2 Molecular Weight (g/mol): 206.29 MDL Number: MFCD00010393 InChI Key: HEFNNWSXXWATRW-UHFFFAOYNA-N Synonym: ibuprofen,2-4-isobutylphenyl propanoic acid,motrin,brufen,nurofen,advil,dolgit,liptan,nuprin,anflagen PubChem CID: 3672 ChEBI: CHEBI:5855 IUPAC Name: 2-[4-(2-methylpropyl)phenyl]propanoic acid SMILES: CC(C)CC1=CC=C(C=C1)C(C)C(O)=O
Ibuprofen , ≥98% (HPLC), MilliporeSigma™ Supelco™
MDL Number: MFCD00010393 Synonym: alpha-Methyl-4-(isobutyl)phenylacetic acid; (+/-)-2-(4-Isobutylphenyl)propanoic acid
Ibuprofen, USP, 97-103%, Spectrum™ Chemical
CAS: 15687-27-1 Molecular Formula: C13H18O2 Molecular Weight (g/mol): 206.29 MDL Number: MFCD00010393 InChI Key: HEFNNWSXXWATRW-UHFFFAOYNA-N IUPAC Name: 2-[4-(2-methylpropyl)phenyl]propanoic acid SMILES: CC(C)CC1=CC=C(C=C1)C(C)C(O)=O
MilliporeSigma™ Calbiochem™ (±)-Ibuprofen
A nonsteroidal anti-inflammatory drug (NSAID) that acts as a reversible and competitive inhibitor of cyclooxygenase 1 (COX-1) (IC50 = 4.85 μM).
4-Isobutyl-alpha-methylphenylacetic acid, 99%
CAS: 15687-27-1 Molecular Formula: C13H18O2 Molecular Weight (g/mol): 206.29 MDL Number: MFCD00010393 InChI Key: HEFNNWSXXWATRW-UHFFFAOYNA-N Synonym: ibuprofen,2-4-isobutylphenyl propanoic acid,motrin,brufen,nurofen,advil,dolgit,liptan,nuprin,anflagen PubChem CID: 3672 ChEBI: CHEBI:5855 IUPAC Name: 2-[4-(2-methylpropyl)phenyl]propanoic acid SMILES: CC(C)CC1=CC=C(C=C1)C(C)C(O)=O
Ibuprofen, 99%, Thermo Scientific Chemicals
CAS: 15687-27-1 Molecular Formula: C13H18O2 Molecular Weight (g/mol): 206.29 MDL Number: MFCD00010393 InChI Key: HEFNNWSXXWATRW-UHFFFAOYNA-N PubChem CID: 3672 ChEBI: CHEBI:5855 IUPAC Name: 2-[4-(2-methylpropyl)phenyl]propanoic acid SMILES: CC(C)CC1=CC=C(C=C1)C(C)C(O)=O
Medisca Ibuprofen USP
Ibuprofen USP is a white to off-white, crystalline powder, having a slight, characteristic odor.