accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1569-02-4

IUPAC Name: 1-ethoxypropan-2-ol
Synonyms: 1-ethoxy-2-propanol propylene glycol monoethyl ether
Molecular Weight (g/mol): 104.15
Molecular Formula: C5H12O2
InChi Key: JOLQKTGDSGKSKJ-UHFFFAOYNA-N
SMILES: CCOCC(C)O
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (7)
  • (7)