accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CPNCH3CH3

CAS RN 157141-27-0

IUPAC Name: 2-(tributyl-λ⁵-phosphanylidene)acetonitrile
Synonyms: 2-(tributyl-λ⁵-phosphanylidene)acetonitrile
Molecular Weight (g/mol): 241.36
Molecular Formula: C14H28NP
InChi Key: OZMLUMPWPFZWTP-UHFFFAOYSA-N
SMILES: CCCCP(CCCC)(CCCC)=CC#N
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)