Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanomethylenetributylphosphorane 97.0+%, TCI America™
SDP

Catalog No. C15005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15005G 5 g
C150025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15005G Supplier TCI America Supplier No. C15005G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 157141-27-0
Molecular Formula C14H28NP
Molecular Weight (g/mol) 241.36
MDL Number MFCD01321151
InChI Key OZMLUMPWPFZWTP-UHFFFAOYSA-N
PubChem CID 4622928
IUPAC Name 2-(tributyl-λ⁵-phosphanylidene)acetonitrile
SMILES CCCCP(CCCC)(CCCC)=CC#N

Specifications

Color Brown-Yellow
Quantity 5 g
Formula Weight 241.36
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Cyanomethylenetributylphosphorane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.