accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3NH3ClTiClClCl

CAS RN 15719-81-0

IUPAC Name: tetrachlorotitanium diamine
Synonyms: titanium tetrachloride diamine
Molecular Weight (g/mol): 223.73
Molecular Formula: Cl4H6N2Ti
InChi Key: OAVZMJMVTGHHAL-UHFFFAOYSA-J
SMILES: N.N.Cl[Ti](Cl)(Cl)Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)