Learn More
Diamminetetrachlorotitanate(IV), 99%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Melting Point | 240°C (decomposition) |
| Boiling Point | 120°C (0.1 mmHg)(sublimation) |
| Quantity | 10 g |
| UN Number | UN3260 |
| Sensitivity | Moisture Sensitive |
| Formula Weight | 223.75 |
| Percent Purity | 99% |
| Physical Form | Powder |
| Chemical Name or Material | Diamminetetrachlorotitanate(IV) |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | GHS P Statement P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a Do not breathe dust/fume/gas/mist/vapors/spray. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.