accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH2NNHNOO

CAS RN 15723-90-7

IUPAC Name: hydrogen 4-nitrobenzene-1-carboximidamide chloride
Synonyms: hydrogen 4-nitrobenzenecarboximidamide chloride
Molecular Weight (g/mol): 201.61
Molecular Formula: C7H8ClN3O2
InChi Key: CJXJAUKCHHAJQN-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC(=N)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results