accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1572936-83-4

IUPAC Name: 2-[(4-methoxynaphthalen-2-yl)amino]-5-nitrobenzoic acid
Synonyms: 2-[(4-methoxynaphthalen-2-yl)amino]-5-nitrobenzoic acid
Molecular Weight (g/mol): 338.32
Molecular Formula: C18H14N2O5
InChi Key: JIVRLHBAUUZTNC-UHFFFAOYSA-N
SMILES: COC1=C2C=CC=CC2=CC(NC2=C(C=C(C=C2)[N+]([O-])=O)C(O)=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results