CAS RN 157408-08-7
CAS RN 157408-08-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bacoside A3, ≥95% (HPLC), MilliporeSigma™ Supelco™
Bacoside A3 is a triterpenoid saponin and an important constituent of bacosides, a saponin mixture of Bacopa monniera.