accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3C

CAS RN 1579-40-4

IUPAC Name: 1-methyl-4-(4-methylphenoxy)benzene
Synonyms: benzene, 1,1'-oxybis 4-methyl-
Molecular Weight (g/mol): 198.27
Molecular Formula: C14H14O
InChi Key: YWYHGNUFMPSTTR-UHFFFAOYSA-N
SMILES: CC1=CC=C(OC2=CC=C(C)C=C2)C=C1
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)