Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-p-tolyl Ether 98.0+%, TCI America™
SDP

Catalog No. D217825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D217825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D217825G Supplier TCI America Supplier No. D217825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1579-40-4
Molecular Formula C14H14O
Molecular Weight (g/mol) 198.265
MDL Number MFCD00025979
InChI Key YWYHGNUFMPSTTR-UHFFFAOYSA-N
Synonym p-tolyl ether, di-p-tolyl ether, 4-tolyl ether, 4,4'-oxybis methylbenzene, bis 4-methylphenyl ether, 4,4'-dimethyldiphenyl ether, benzene, 1,1'-oxybis 4-methyl, p-p-tolyloxy toluene, 1,1'-oxybis 4-methylbenzene
PubChem CID 74098
IUPAC Name 1-methyl-4-(4-methylphenoxy)benzene
SMILES CC1=CC=C(C=C1)OC2=CC=C(C=C2)C

Specifications

Melting Point 48°C
Color White
Boiling Point 285°C
Quantity 25 g
Formula Weight 198.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Di-p-tolyl Ether
EINECSNumber (3)-0653
RTECSNumber DA6625000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.