accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONFFFFF

CAS RN 1580464-65-8

IUPAC Name: 1-[6-(1,1,2,2,2-pentafluoroethyl)pyridin-2-yl]ethan-1-one
Synonyms: 1-[6-(1,1,2,2,2-pentafluoroethyl)pyridin-2-yl]ethanone
Molecular Weight (g/mol): 239.15
Molecular Formula: C9H6F5NO
InChi Key: KCNUAVPBVPTCLU-UHFFFAOYSA-N
SMILES: CC(=O)C1=NC(=CC=C1)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results