accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONNOOMn(?)(E/Z)(E/Z)(?)

CAS RN 158052-18-7

IUPAC Name: manganese(3+) 3-[({2-[({2-oxido-2'-phenyl-[1,1'-binaphthalen]-3-yl}methylidene)amino]-1,2-diphenylethyl}imino)methyl]-2'-phenyl-[1,1'-binaphthalen]-2-olate acetate
Synonyms: manganese(3+) 3-[({2-[({2-oxido-2'-phenyl-[1,1'-binaphthalen]-3-yl}methylidene)amino]-1,2-diphenylethyl}imino)methyl]-2'-phenyl-[1,1'-binaphthalen]-2-olate acetate
Molecular Weight (g/mol): 1037.11
Molecular Formula: C70H49MnN2O4
InChi Key: KXEXRGSEABDUME-UHFFFAOYNA-K
SMILES: [Mn+3].CC([O-])=O.[O-]C1=C(C=NC(C(N=CC2=C([O-])C(=C3C=CC=CC3=C2)C2=C3C=CC=CC3=CC=C2C2=CC=CC=C2)C2=CC=CC=C2)C2=CC=CC=C2)C=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results