Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-Acetamido-7,8, 9-tri-O-acetyl-5-N,4-O-carbonyl-3,5-dideoxy-2-S-phenyl-2-thio-D-glycero-β 89.0+%, TCI America™
SDP

Catalog No. B2409100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2409100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B2409100MG Supplier TCI America Supplier No. B2409100MG

Chemical Identifiers

CAS 158052-18-7
Molecular Formula C70H49MnN2O4
Molecular Weight (g/mol) 1037.11
MDL Number MFCD06797076
InChI Key KXEXRGSEABDUME-XKCBYAASSA-K
PubChem CID 53384488
IUPAC Name manganese(3+);3-[[(1S,2S)-2-[[3-oxido-4-(2-phenylnaphthalen-1-yl)naphthalen-2-yl]methylideneamino]-1,2-diphenylethyl]iminomethyl]-1-(2-phenylnaphthalen-1-yl)naphthalen-2-olate;acetate
SMILES CC(=O)[O-].C1=CC=C(C=C1)C2=C(C3=CC=CC=C3C=C2)C4=C(C(=CC5=CC=CC=C54)C=NC(C6=CC=CC=C6)C(C7=CC=CC=C7)N=CC8=CC9=CC=CC=C9C(=C8[O-])C1=C(C=CC2=CC=CC=C21)C1=CC=CC=C1)[O-].[Mn+3]

Specifications

Quantity 100 mg
Formula Weight 1037.11
Percent Purity ≥89.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (1S,2S)-N,N′-Bis[(R)-2-hydroxy-2′-phenyl-1,1′-binaphthyl-3-ylmethylene]-1,2-diphen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.