accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOHOOOOOOOOOO

CAS RN 15826-37-6

IUPAC Name: disodium 5-{3-[(2-carboxylato-4-oxo-4H-chromen-5-yl)oxy]-2-hydroxypropoxy}-4-oxo-4H-chromene-2-carboxylate
Synonyms: sodium cromoglycate sodium cromoglicate sodium cromolyn cromoglycate disodium disodium cromoglycate natrium cromoglicat
Molecular Weight (g/mol): 512.33
Molecular Formula: C23H14Na2O11
InChi Key: VLARUOGDXDTHEH-UHFFFAOYSA-L
SMILES: [Na+].[Na+].OC(COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O)COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (8)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
Grade 
  • (6)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (8)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (6)
  • (2)
  • (3)