accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15862-30-3

IUPAC Name: 3-bromo-5-nitropyridine
Synonyms: 3-bromo-5-nitropyridine
Molecular Weight (g/mol): 203
Molecular Formula: C5H3BrN2O2
InChi Key: WHYJVHQKKJHXTB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Br)=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results