CAS RN 158704-02-0
IUPAC Name:
5H,6H,6aH,7H,8H,12bH-benzo[a]phenanthridine-10,11-diol hydrochloride
Synonyms:
dihydrexidine hydrochloride
Molecular Weight (g/mol):
303.79
Molecular Formula:
C17H18ClNO2
InChi Key:
IJYUPBNUPIRQEP-UHFFFAOYNA-N
SMILES:
Cl.OC1=C(O)C=C2C3C(CCC2=C1)NCC1=CC=CC=C31
1
–
1
of
1
results
1
R&D Systems™ Dihydrexidine hydrochloride
CAS: 158704-02-0 Molecular Formula: C17H18ClNO2