accessibility menu, dialog, popup

UserName

Viewing profile as user:
CHFFF

CAS RN 158816-55-8

IUPAC Name: 5-ethynyl-1,2,3-trifluorobenzene
Synonyms: 5-ethynyl-1,2,3-trifluorobenzene
Molecular Weight (g/mol): 156.11
Molecular Formula: C8H3F3
InChi Key: BZXWRVPVZZZAKB-UHFFFAOYSA-N
SMILES: FC1=CC(=CC(F)=C1F)C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results