accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15901-42-5

IUPAC Name: 3,3,5-trimethylcyclohexan-1-amine
Synonyms: 3,3,5-trimethylcyclohexan-1-amine
Molecular Weight (g/mol): 141.26
Molecular Formula: C9H19N
InChi Key: ZGMQLPDXPUINCQ-UHFFFAOYNA-N
SMILES: CC1CC(N)CC(C)(C)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results