accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOSOHOO

CAS RN 15909-83-8

IUPAC Name: 3-hydroxypropane-1-sulfonic acid
Synonyms: 3-hydroxypropane-1-sulfonic acid
Molecular Weight (g/mol): 140.15
Molecular Formula: C3H8O4S
InChi Key: WQPMYSHJKXVTME-UHFFFAOYSA-N
SMILES: OCCCS(O)(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results