accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO(R)SNHOO

CAS RN 159453-24-4

IUPAC Name: (2R)-2-{[(benzyloxy)carbonyl]amino}-3-(phenylsulfanyl)propanoate
Synonyms: (2R)-2-{[(benzyloxy)carbonyl]amino}-3-(phenylsulfanyl)propanoate
Molecular Weight (g/mol): 330.38
Molecular Formula: C17H16NO4S
InChi Key: ISBOGFMUFMJWEP-HNNXBMFYSA-M
SMILES: [O-]C(=O)[C@H](CSC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results