CAS RN 159533-25-2
IUPAC Name:
3-[3-(dimethylamino)propyl]-4-hydroxy-N-[4-(pyridin-4-yl)phenyl]benzamide dihydrochloride
Synonyms:
3-[3-(dimethylamino)propyl]-4-hydroxy-N-[4-(pyridin-4-yl)phenyl]benzamide dihydrochloride
Molecular Weight (g/mol):
448.39
Molecular Formula:
C23H27Cl2N3O2
InChi Key:
KBKWJHYQFQONBJ-UHFFFAOYSA-N
SMILES:
Cl.Cl.CN(C)CCCC1=C(O)C=CC(=C1)C(=O)NC1=CC=C(C=C1)C1=CC=NC=C1
1
–
1
of
1
results
1
R&D Systems™ GR 55562 dihydrochloride
CAS: 159533-25-2 Molecular Formula: C23H27Cl2N3O2