accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15971-29-6

CH3OO

CAS RN 15971-29-6

IUPAC Name: 4-methoxynaphthalene-1-carbaldehyde
Synonyms: 4-methoxynaphthalene-1-carbaldehyde
Molecular Weight (g/mol): 186.21
Molecular Formula: C12H10O2
InChi Key: MVXMNHYVCLMLDD-UHFFFAOYSA-N
SMILES: COC1=C2C=CC=CC2=C(C=O)C=C1
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)