Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-1-naphthaldehyde 97.0+%, TCI America™
SDP

Catalog No. M10495G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M10495G Supplier TCI America Supplier No. M10495G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15971-29-6
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00004006
InChI Key MVXMNHYVCLMLDD-UHFFFAOYSA-N
Synonym 4-methoxy-1-naphthaldehyde, 4-methoxy-1-naphthalaldehyde, 1-naphthalenecarboxaldehyde, 4-methoxy, 4-methoxynaphthalenecarbaldehyde, pubchem16658, acmc-209dk5, ksc494i2d, 1-naphthaldehyde, 4-methoxy, 4-methoxy-1-naphthalenecarbaldehyde
PubChem CID 85217
IUPAC Name 4-methoxynaphthalene-1-carbaldehyde
SMILES COC1=CC=C(C2=CC=CC=C21)C=O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 186.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Methoxy-1-naphthaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.