accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNONNOOOCl

CAS RN 159717-68-7

IUPAC Name: 1-(7-nitro-2,1,3-benzoxadiazol-4-yl)pyrrolidine-2-carbonyl chloride
Synonyms: 1-(7-nitro-2,1,3-benzoxadiazol-4-yl)pyrrolidine-2-carbonyl chloride
Molecular Weight (g/mol): 296.67
Molecular Formula: C11H9ClN4O4
InChi Key: XBQYXPYABMCADZ-UHFFFAOYNA-N
SMILES: [O-][N+](=O)C1=CC=C(N2CCCC2C(Cl)=O)C2=NON=C12
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results