accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OOHH3CH3C(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 160162-42-5

IUPAC Name: 3-methyl-7-(4-methylphenyl)-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid
Synonyms: 3-methyl-7-(4-methylphenyl)-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid
Molecular Weight (g/mol): 376.54
Molecular Formula: C26H32O2
InChi Key: RQANARBNMTXCDM-UHFFFAOYSA-N
SMILES: CC(C=CC=C(C=CC1=C(C)CCCC1(C)C)C1=CC=C(C)C=C1)=CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results