accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1606-08-2

IUPAC Name: cyclopentylcyclohexane
Synonyms: cyclopentylcyclohexane
Molecular Weight (g/mol): 152.28
Molecular Formula: C11H20
InChi Key: WVABZRMBCQFXBK-UHFFFAOYSA-N
SMILES: C1CCC(C1)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results