accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1606-85-5

IUPAC Name: 2-{[4-(2-hydroxyethoxy)but-2-yn-1-yl]oxy}ethan-1-ol
Synonyms: 2-{[4-(2-hydroxyethoxy)but-2-yn-1-yl]oxy}ethanol
Molecular Weight (g/mol): 174.2
Molecular Formula: C8H14O4
InChi Key: IXAWTPMDMPUGLV-UHFFFAOYSA-N
SMILES: OCCOCC#CCOCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results