Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis(2-hydroxyethoxy)-2-butyne 50.0+%, TCI America™
SDP

Catalog No. B097925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B097925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B097925G Supplier TCI America Supplier No. B097925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1606-85-5
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD00002883
InChI Key IXAWTPMDMPUGLV-UHFFFAOYSA-N
Synonym 1,4-bis 2-hydroxyethoxy-2-butyne, 2-4-2-hydroxyethoxy but-2-ynoxy ethanol, unii-8tt85143cr, 2,2'-but-2-yne-1,4-diylbis oxy diethanol, ethanol, 2,2'-2-butyne-1,4-diylbis oxy bis, 2,2'-but-2-ynylenedioxydiethanol, ethanol, 2,2'-2-butynylenedioxy di, 2,2'-2-butyne-1,4-diyldioxy diethanol, 2,2'-2-butyne-1,4-diylbis oxy bisethanol, 3,8-dioxa-5-decyne-1,10-diol
PubChem CID 62411
IUPAC Name 2-[4-(2-hydroxyethoxy)but-2-ynoxy]ethanol
SMILES C(COCC#CCOCCO)O

Specifications

Quantity 25 g
Formula Weight 174.20
Percent Purity ≥50.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Bis(2-hydroxyethoxy)-2-butyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.