accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1608-41-9

IUPAC Name: 1,4-bis(2-phenylethenyl)benzene
Synonyms: 1,4-bis(2-phenylethenyl)benzene
Molecular Weight (g/mol): 282.39
Molecular Formula: C22H18
InChi Key: IJAAWBHHXIWAHM-UHFFFAOYSA-N
SMILES: C(=CC1=CC=C(C=CC2=CC=CC=C2)C=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)