Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis(trans-2-phenylethenyl)benzene, 97%, Thermo Scientific™

Catalog No. AAH3143603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3143603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3143603 Supplier Thermo Scientific Chemicals Supplier No. H3143603

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1608-41-9
Molecular Formula C22H18
Molecular Weight (g/mol) 282.386
MDL Number MFCD00154950
InChI Key IJAAWBHHXIWAHM-PHEQNACWSA-N
Synonym 1,4-distyrylbenzene, 1,4-bis e-2-phenylethenyl benzene, trans,trans-1,4-distyrylbenzene, 1,4-bis trans-2-phenylethenyl benzene, benzene, 1,4-bis 2-phenylethenyl, benzene, 1,4-bis 1e-2-phenylethenyl, 1,4-bis trans-styryl benzene, 1,4-bis e-styryl benzene, 4-e-styryl-cis-stilbene, 1,4-di trans-beta-styryl benzene
PubChem CID 640299
IUPAC Name 1,4-bis[(E)-2-phenylethenyl]benzene
SMILES C1=CC=C(C=C1)C=CC2=CC=C(C=C2)C=CC3=CC=CC=C3

Specifications

Melting Point 264°C to 266°C
Quantity 1 g
Formula Weight 282.39
Percent Purity 97%
Chemical Name or Material 1,4-Bis(trans-2-phenylethenyl)benzene
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.