Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-1,4-Distyrylbenzene 98.0+%, TCI America™
SDP

Catalog No. D42991G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D42991G Supplier TCI America Supplier No. D42991G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1608-41-9
Molecular Formula C22H18
Molecular Weight (g/mol) 282.386
MDL Number MFCD00154950
InChI Key IJAAWBHHXIWAHM-PHEQNACWSA-N
Synonym 1,4-distyrylbenzene, 1,4-bis e-2-phenylethenyl benzene, trans,trans-1,4-distyrylbenzene, 1,4-bis trans-2-phenylethenyl benzene, benzene, 1,4-bis 2-phenylethenyl, benzene, 1,4-bis 1e-2-phenylethenyl, 1,4-bis trans-styryl benzene, 1,4-bis e-styryl benzene, 4-e-styryl-cis-stilbene, 1,4-di trans-beta-styryl benzene
PubChem CID 640299
IUPAC Name 1,4-bis[(E)-2-phenylethenyl]benzene
SMILES C1=CC=C(C=C1)C=CC2=CC=C(C=C2)C=CC3=CC=CC=C3

Specifications

Melting Point 266°C
Quantity 1 g
Formula Weight 282.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-1,4-Distyrylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.