accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 160848-22-6

IUPAC Name: methyl 4-{54-phenyldotriacontacyclo[19.16.16.3²,⁷.2¹³,¹⁵.1⁴,⁴².1⁵,⁹.1¹⁶,³⁷.0³,³⁹.0⁴,⁴².0⁸,¹².0¹⁰,⁴⁵.0¹¹,⁴⁸.0¹⁷,³⁶.0¹⁸,²².0²⁰,⁵⁰.0²³,³⁵.0²⁴,³³.0²⁵,⁵³.0²⁶,³¹.0²⁷,⁵².0²⁸,⁴⁶.0²⁹,⁴⁴.0³⁰,⁴¹.0³²,⁴⁰.0³⁴,³⁸.0⁴⁷,⁵¹.0⁶,⁶¹.0¹⁴,⁵⁹.0¹⁹,⁵⁶.0⁴³,⁵⁸.0⁴⁹,⁵⁷.0⁵⁵,⁶⁰]henhexaconta-1,3(39),5(58),6(61),7,9,11,13,15(56),16(55),17,19,21,23(35),24(33),25(53),26(31),27(52),28,30(41),32(40),34(38),36,43,45,47,49(57),50,59-nonacosaen-54-yl}butanoate
Synonyms: methyl 4-{54-phenyldotriacontacyclo[19.16.16.3²,⁷.2¹³,¹⁵.1⁴,⁴².1⁵,⁹.1¹⁶,³⁷.0³,³⁹.0⁴,⁴².0⁸,¹².0¹⁰,⁴⁵.0¹¹,⁴⁸.0¹⁷,³⁶.0¹⁸,²².0²⁰,⁵⁰.0²³,³⁵.0²⁴,³³.0²⁵,⁵³.0²⁶,³¹.0²⁷,⁵².0²⁸,⁴⁶.0²⁹,⁴⁴.0³⁰,⁴¹.0³²,⁴⁰.0³⁴,³⁸.0⁴⁷,⁵¹.0⁶,⁶¹.0¹⁴,⁵⁹.0¹⁹,⁵⁶.0⁴³,⁵⁸.0⁴⁹,⁵⁷.0⁵⁵,⁶⁰]henhexaconta-1,3(39),5(58),6(61),7,9,11,13,15(56),16(55),17,19,21,23(35),24(33),25(53),26(31),27(52),28,30(41),32(40),34(38),36,43,45,47,49(57),50,59-nonacosaen-54-yl}butanoate
Molecular Weight (g/mol): 910.9
Molecular Formula: C72H14O2
InChi Key: MCEWYIDBDVPMES-UHFFFAOYNA-N
SMILES: COC(=O)CCCC1(C2=CC=CC=C2)C23C4=C5C6=C7C8=C9C%10=C%11C%12=C%13C%14=C%15C%16=C%17C(=C8C%16=C%10%13)C6=C4C4=C6C8=C%10C%13=C%16C%18=C%19C(=C%12C%18=C%14C%13=C%15C8=C%174)C4=C8C%12=C%13C(C7=C9C%12=C%114)=C5C4=C%13C5=C8C%19=C7C%16=C%10C(=C26)C(=C57)C134
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (3)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)