accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16096-33-6

IUPAC Name: 1-phenyl-1H-indole
Synonyms: 1-phenylindole
Molecular Weight (g/mol): 193.25
Molecular Formula: C14H11N
InChi Key: YBFCBQMICVOSRW-UHFFFAOYSA-N
SMILES: C1=CC2=CC=CC=C2N1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results