Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1H-indole 98.0+%, TCI America™
SDP

Catalog No. P25031G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P25031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P25031G Supplier TCI America Supplier No. P25031G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16096-33-6
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD00123280
InChI Key YBFCBQMICVOSRW-UHFFFAOYSA-N
PubChem CID 182029
IUPAC Name 1-phenylindole
SMILES C1=CC=C(C=C1)N2C=CC3=CC=CC=C32

Specifications

Color Yellow
Boiling Point 190°C
Quantity 1 g
Formula Weight 193.25
Percent Purity ≥98.0% (N)
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1H-indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.