accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNClNFFF

CAS RN 16097-63-5

IUPAC Name: 4-chloro-2-(methylsulfanyl)-6-(trifluoromethyl)pyrimidine
Synonyms: 4-chloro-2-(methylsulfanyl)-6-(trifluoromethyl)pyrimidine
Molecular Weight (g/mol): 228.62
Molecular Formula: C6H4ClF3N2S
InChi Key: GOORQLLDZBAQIW-UHFFFAOYSA-N
SMILES: CSC1=NC(Cl)=CC(=N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results