accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 161058-83-9

IUPAC Name: 6-(cyclohexylmethoxy)-7H-purin-2-amine
Synonyms: 6-(cyclohexylmethoxy)-7H-purin-2-amine
Molecular Weight (g/mol): 247.3
Molecular Formula: C12H17N5O
InChi Key: MWGXGTJJAOZBNW-UHFFFAOYSA-N
SMILES: NC1=NC(OCC2CCCCC2)=C2NC=NC2=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Product Line 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)