Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NU 2058 98.0+%, TCI America™
SDP

Catalog No. N095820MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N095820MG 20 mg
N0958100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N095820MG Supplier TCI America Supplier No. N095820MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 161058-83-9
Molecular Formula C12H17N5O
Molecular Weight (g/mol) 247.302
MDL Number MFCD05664734
InChI Key MWGXGTJJAOZBNW-UHFFFAOYSA-N
Synonym 6-cyclohexylmethoxy-9h-purin-2-amine, 6-o-cyclohexylmethyl guanine, 2-amino-6-cyclohexylmethoxypurine, 2-amino-6-cyclohexylmethyl oxy purine, 6-cyclohexylmethoxy-7h-purin-2-amine, 9h-purin-2-amine, 6-cyclohexylmethoxy, o-cyclohexylmethylguanine, o6-cyclohexylmethylguanine, d0r1ft, d0sj6y
PubChem CID 4564
IUPAC Name 6-(cyclohexylmethoxy)-7H-purin-2-amine
SMILES C1CCC(CC1)COC2=NC(=NC3=C2NC=N3)N

Specifications

Melting Point 199°C
Color White-Yellow
Quantity 20 mg
Formula Weight 247.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material NU 2058
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.